accessibility menu, dialog, popup

UserName

CAS RN 446-17-3

OOFFF

CAS RN 446-17-3

IUPAC Name: 2,4,5-trifluorobenzoate
Synonyms: 2,4,5-trifluorobenzoate
Molecular Weight (g/mol): 175.09
Molecular Formula: C7H2F3O2
InChi Key: AKAMNXFLKYKFOJ-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC(F)=C(F)C=C1F
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results