accessibility menu, dialog, popup

UserName

CAS RN 4314-66-3

H3CHNON

CAS RN 4314-66-3

IUPAC Name: N-ethylpyridine-3-carboxamide
Synonyms: N-ethylnicotinamide
Molecular Weight (g/mol): 150.18
Molecular Formula: C8H10N2O
InChi Key: ZXOAHASJYIUCBG-UHFFFAOYSA-N
SMILES: CCNC(=O)C1=CC=CN=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results