accessibility menu, dialog, popup

UserName

CAS RN 43023-95-6

FAsFFFFFRb

CAS RN 43023-95-6

IUPAC Name: rubidium(1+) hexafluoro-λ⁵-arsanuide
Synonyms: rubidium(1+) hexafluoro-λ⁵-arsanuide
Molecular Weight (g/mol): 274.38
Molecular Formula: AsF6Rb
InChi Key: FDUSNUSHTOTAKN-UHFFFAOYSA-N
SMILES: [Rb+].F[As-](F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results