CAS RN 4295-06-1
CAS RN 4295-06-1
IUPAC Name:
4-chloro-2-methylquinoline
Synonyms:
4-chloro-2-methyl-quinoline
Molecular Weight (g/mol):
177.63
Molecular Formula:
C10H8ClN
InChi Key:
HQAIROMRVBVWSK-UHFFFAOYSA-N
SMILES:
CC1=CC(Cl)=C2C=CC=CC2=N1
1
–
2
of
2
results
1
4-Chloro-2-methylquinoline, 97%
CAS: 4295-06-1 MDL Number: MFCD00006757
2
4-Chloroquinaldine, 99%
CAS: 4295-06-1 Molecular Formula: C10H8ClN Molecular Weight (g/mol): 177.63 InChI Key: HQAIROMRVBVWSK-UHFFFAOYSA-N PubChem CID: 77973 IUPAC Name: 4-chloro-2-methylquinoline SMILES: CC1=NC2=CC=CC=C2C(=C1)Cl