CAS RN 424818-25-7
CAS RN 424818-25-7
IUPAC Name:
N-tert-butyl-3-nitrobenzene-1-sulfonamide
Synonyms:
N-tert-butyl-3-nitrobenzenesulfonamide
Molecular Weight (g/mol):
258.29
Molecular Formula:
C10H14N2O4S
InChi Key:
UHFXZEXYBBMXBY-UHFFFAOYSA-N
SMILES:
CC(C)(C)NS(=O)(=O)C1=CC=CC(=C1)[N+]([O-])=O
1
–
3
of
3
results
1
N-tert-Butyl 3-Nitrobenzenesulfonamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-tert-Butyl 3-Nitrobenzenesulfonamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-tert-Butyl 3-Nitrobenzenesulfonamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.