CAS RN 4197-24-4
CAS RN 4197-24-4
IUPAC Name:
hydrogen 4-[(4-aminophenyl)[(1Z)-4-imino-3-methylcyclohexa-2,5-dien-1-ylidene]methyl]-2-methylaniline chloride
Synonyms:
hydrogen 4-[(4-aminophenyl)[(1Z)-4-imino-3-methylcyclohexa-2,5-dien-1-ylidene]methyl]-2-methylaniline chloride
Molecular Weight (g/mol):
351.88
Molecular Formula:
C21H22ClN3
InChi Key:
HZLHRDBTVSZCBS-GHTYLULLSA-N
SMILES:
[H+].[Cl-].CC1=CC(=CC=C1N)C(\C1=CC=C(N)C=C1)=C1\C=CC(=N)C(C)=C1
1
–
2
of
2
results
1
Thermo Scientific Chemicals Carbol Fuchsin, pure
CAS: 4197-24-4 Molecular Formula: C21H22ClN3 Molecular Weight (g/mol): 351.878 MDL Number: MFCD00143923 InChI Key: HZLHRDBTVSZCBS-GHTYLULLSA-N PubChem CID: 91997463 IUPAC Name: 4-[(Z)-(4-aminophenyl)-(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-2-methylaniline;hydron;chloride SMILES: [H+].CC1=CC(=C(C2=CC=C(C=C2)N)C3=CC(=C(C=C3)N)C)C=CC1=N.[Cl-]