accessibility menu, dialog, popup

UserName

CAS RN 4196-99-0

NaNaOSOONNHNNOSOOO(E/Z)(Z)

CAS RN 4196-99-0

IUPAC Name: disodium 2-{2-[(1Z)-2-oxo-1,2-dihydronaphthalen-1-ylidene]hydrazin-1-yl}-5-[2-(4-sulfonatophenyl)diazen-1-yl]benzene-1-sulfonate
Synonyms: biebrich scarlet
Molecular Weight (g/mol): 556.47
Molecular Formula: C22H14N4Na2O7S2
InChi Key: YCKMEQRBEVVZQF-VKZXTWAVSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)N=NC1=CC=C(N\N=C2/C(=O)C=CC3=CC=CC=C23)C(=C1)S([O-])(=O)=O
Quantity 
  • (2)
  • (1)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)