CAS RN 4091-39-8
CAS RN 4091-39-8
IUPAC Name:
3-chlorobutan-2-one
Synonyms:
3-chloro-2-butanone
Molecular Weight (g/mol):
106.55
Molecular Formula:
C4H7ClO
InChi Key:
OIMRLHCSLQUXLL-UHFFFAOYNA-N
SMILES:
CC(Cl)C(C)=O
1
–
1
of
1
results
1
3-Chloro-2-butanone, 96%
CAS: 4091-39-8 Molecular Formula: C4H7ClO Molecular Weight (g/mol): 106.549 MDL Number: MFCD00000865 InChI Key: OIMRLHCSLQUXLL-UHFFFAOYSA-N Synonym: 3-chloro-2-butanone,2-butanone, 3-chloro,3-chlorobutanone,1-acetyl-1-chloroethane,1-chloroethyl methyl ketone,2-chloro-3-butanone,2-butanone, 3-chloro-, +,chloro-mek,3-chloro-2-butanon,3-chlorobutan-2-on PubChem CID: 20026 IUPAC Name: 3-chlorobutan-2-one SMILES: CC(C(=O)C)Cl