accessibility menu, dialog, popup

UserName

CAS RN 4073-98-7

CH3CH3NH2H3CH3CH2N

CAS RN 4073-98-7

IUPAC Name: 4-[(4-amino-3,5-dimethylphenyl)methyl]-2,6-dimethylaniline
Synonyms: 4-[(4-amino-3,5-dimethylphenyl)methyl]-2,6-dimethylaniline
Molecular Weight (g/mol): 254.38
Molecular Formula: C17H22N2
InChi Key: OMHOXRVODFQGCA-UHFFFAOYSA-N
SMILES: CC1=CC(CC2=CC(C)=C(N)C(C)=C2)=CC(C)=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results