CAS RN 3999-38-0
IUPAC Name:
2-methylpyridine boron
Synonyms:
α-methylpyridine boron
Molecular Weight (g/mol):
103.94
Molecular Formula:
C6H7BN
InChi Key:
QHXLIQMGIGEHJP-UHFFFAOYSA-N
SMILES:
[B].CC1=CC=CC=N1
1
–
3
of
3
results
1
Borane-2-methylpyridine complex, 95%
CAS: 3999-38-0 Molecular Formula: C6H10BN Molecular Weight (g/mol): 106.96 MDL Number: MFCD07784361 InChI Key: ZMASBEFBIXMNCP-UHFFFAOYSA-N Synonym: 2-Picoline borane complex IUPAC Name: 2-methylpyridine borane SMILES: B.CC1=CC=CC=N1
2
Borane-2-methylpyridine Complex, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Borane-2-methylpyridine Complex, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.