CAS RN 3947-65-7
CAS RN 3947-65-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Neamine, TRC
CAS: 3947-65-7 Molecular Formula: C12 H26 N4 O6 Molecular Weight (g/mol): 322.36 Synonym: 2-Deoxy-4-O-(2,6-diamino-2,6-dideoxy-α-D-glucopyranosyl)-D-streptamine,Neomycin A (6CI,7CI,8CI),(+)-Neamine,Dekamycin V,Neamin,Neamine,Nebramycin X,Negamicin,O-2,6-Diamino-2,6-dideoxy-α-D-glucopyranosyl-(1' ' ’4)-1,3-diamino-1,2,3-trideoxy-D-myo-inositol,ST 7,Neomycin Imp. A (EP),GW 318764X IUPAC Name: (2R,3S,4R,5R,6R)-5-amino-2-(aminomethyl)-6-[(1R,2R,3S,4R,6S)-4,6-diamino-2,3-dihydroxycyclohexyl]oxyoxane-3,4-diol SMILES: NC[C@H]1O[C@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O)[C@H]2O)[C@H](N)[C@@H](O)[C@@H]1O