CAS RN 39430-51-8
CAS RN 39430-51-8
IUPAC Name:
chromium(3+) triacetate
Synonyms:
chromium acetate hydroxide
Molecular Weight (g/mol):
229.13
Molecular Formula:
C6H9CrO6
InChi Key:
WYYQVWLEPYFFLP-UHFFFAOYSA-K
SMILES:
[Cr+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
1
–
1
of
1
results
1
Chromium(III) acetate hydroxide, Cr 24%
CAS: 39430-51-8 Molecular Formula: Cr3(OH)2(OOCCH3)7 MDL Number: MFCD00058912