CAS RN 392326-81-7
CAS RN 392326-81-7
IUPAC Name:
4-(4-bromophenyl)pyrimidin-2-amine
Synonyms:
4-(4-bromophenyl)pyrimidin-2-amine
Molecular Weight (g/mol):
250.1
Molecular Formula:
C10H8BrN3
InChi Key:
ULMOYCJVOJTPCJ-UHFFFAOYSA-N
SMILES:
NC1=NC=CC(=N1)C1=CC=C(Br)C=C1
1
–
1
of
1
results
1
2-Amino-4-(4-bromophenyl)pyrimidine, 98%, Thermo Scientific™
CAS: 392326-81-7 Molecular Formula: C10H8BrN3 Molecular Weight (g/mol): 250.099 MDL Number: MFCD02317272 InChI Key: ULMOYCJVOJTPCJ-UHFFFAOYSA-N Synonym: 4-4-bromophenyl pyrimidin-2-amine,2-amino-4-4-bromophenyl pyrimidine,2-pyrimidinamine,4-4-bromophenyl,4-4-bromophenyl-2-pyrimidinylamine,2-pyrimidinamine, 4-4-bromophenyl,4-4-bromo-phenyl-pyrimidin-2-ylamine PubChem CID: 4590150 IUPAC Name: 4-(4-bromophenyl)pyrimidin-2-amine SMILES: C1=CC(=CC=C1C2=NC(=NC=C2)N)Br