CAS RN 3910-58-5
CAS RN 3910-58-5
IUPAC Name:
butyl[(4-methoxyphenyl)methyl]amine
Synonyms:
butyl[(4-methoxyphenyl)methyl]amine
Molecular Weight (g/mol):
193.29
Molecular Formula:
C12H19NO
InChi Key:
IBTNQSONKQDXNY-UHFFFAOYSA-N
SMILES:
CCCCNCC1=CC=C(OC)C=C1
1
–
2
of
2
results
1
N-Butyl-p-methoxy-benzylamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-Butyl-p-methoxy-benzylamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.