accessibility menu, dialog, popup

UserName

CAS RN 3900-93-4

HOO

CAS RN 3900-93-4

IUPAC Name: 2-cyclopentyl-2-phenylacetic acid
Synonyms: cyclopentyl(phenyl)acetic acid
Molecular Weight (g/mol): 204.27
Molecular Formula: C13H16O2
InChi Key: BCJIDGDYYYBNNB-UHFFFAOYNA-N
SMILES: OC(=O)C(C1CCCC1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results