CAS RN 38677-66-6
CAS RN 38677-66-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Oleoyl-L-carnitine, TRC
CAS: 38677-66-6 Molecular Formula: C25 H47 N O4 Molecular Weight (g/mol): 425.64 Synonym: 1-Propanaminium, 3-carboxy-N,N,N-trimethyl-2-[[(9Z)-1-oxo-9-octadecen-1-yl]oxy]-, inner salt, (2R)-,1-Propanaminium, 3-carboxy-N,N,N-trimethyl-2-[(1-oxo-9-octadecenyl)oxy]-, inner salt, [R-(Z)]-,1-Propanaminium, 3-carboxy-N,N,N-trimethyl-2-[[(9Z)-1-oxo-9-octadecenyl]oxy]-, inner salt, (2R)- (9CI),C18:1 Carnitine,Oleoyl-L-carnitine,Oleoyl L-carnitine,Oleoylcarnitine IUPAC Name: (3R)-3-[(Z)-octadec-9-enoyl]oxy-4-(trimethylazaniumyl)butanoate SMILES: CCCCCCCC\C=C/CCCCCCCC(=O)O[C@H](CC(=O)[O-])C[N+](C)(C)C