CAS RN 38484-94-5
CAS RN 38484-94-5
IUPAC Name:
2,5-dichlorobenzene-1-sulfonic acid dihydrate
Synonyms:
2,5-dichlorobenzenesulfonic acid dihydrate
Molecular Weight (g/mol):
263.09
Molecular Formula:
C6H8Cl2O5S
InChi Key:
VPJICTWHRNQYRG-UHFFFAOYSA-N
SMILES:
O.O.OS(=O)(=O)C1=C(Cl)C=CC(Cl)=C1
1
–
1
of
1
results
1
2,5-Dichlorobenzenesulfonic acid dihydrate, 97%, Thermo Scientific™
CAS: 38484-94-5 Molecular Formula: C6H8Cl2O5S Molecular Weight (g/mol): 263.085 MDL Number: MFCD08271971 InChI Key: VPJICTWHRNQYRG-UHFFFAOYSA-N Synonym: 2,5-dichlorobenzenesulfonic acid dihydrate,acmc-209qt1,2,5-dichlorobenzene-1-sulfonic acid-water 1/2 PubChem CID: 21114734 IUPAC Name: 2,5-dichlorobenzenesulfonic acid;dihydrate SMILES: C1=CC(=C(C=C1Cl)S(=O)(=O)O)Cl.O.O