accessibility menu, dialog, popup

UserName

CAS RN 38220-75-6

H3COOBr

CAS RN 38220-75-6

IUPAC Name: 1-(5-bromo-1-benzofuran-2-yl)ethan-1-one
Synonyms: 1-(5-bromo-1-benzofuran-2-yl)ethanone
Molecular Weight (g/mol): 239.07
Molecular Formula: C10H7BrO2
InChi Key: YPIQPBGNNXXKNI-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC2=CC(Br)=CC=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results