accessibility menu, dialog, popup

UserName

CAS RN 37199-66-9

SKKn

CAS RN 37199-66-9

IUPAC Name: potassium sulfane
Synonyms: potassium hydrosulfide
Molecular Weight (g/mol): 73.17
Molecular Formula: H2KS
InChi Key: ZOCLAPYLSUCOGI-UHFFFAOYSA-N
SMILES: S.[K+]

 No Refinements