accessibility menu, dialog, popup

UserName

CAS RN 36476-87-6

NOHO

CAS RN 36476-87-6

IUPAC Name: 1-(diphenylmethyl)azetidine-3-carboxylic acid
Synonyms: 1-(diphenylmethyl)azetidine-3-carboxylic acid
Molecular Weight (g/mol): 267.33
Molecular Formula: C17H17NO2
InChi Key: BRSCYENHLCPOAU-UHFFFAOYSA-N
SMILES: OC(=O)C1CN(C1)C(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results