accessibility menu, dialog, popup

UserName

CAS RN 3567-62-2

H3CNHONHClCl

CAS RN 3567-62-2

IUPAC Name: 1-(3,4-dichlorophenyl)-3-methylurea
Synonyms: 1-(3,4-dichlorophenyl)-3-methylurea
Molecular Weight (g/mol): 219.07
Molecular Formula: C8H8Cl2N2O
InChi Key: IDQHRQQSSQDLTR-UHFFFAOYSA-N
SMILES: CNC(=O)NC1=CC=C(Cl)C(Cl)=C1
Concentration 
  • (2)
  • (2)
Solvent or Matrix 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)