CAS RN 3565-26-2
CAS RN 3565-26-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-Nitroso-quinolin-8-ol, TRC
CAS: 3565-26-2 Molecular Formula: C9H6N2O2 Molecular Weight (g/mol): 174.16 Synonym: 5-Nitroso-8-quinolinol,5-Nitroso-8-hydroxyquinoline,5-Nitrosooxine,Hydron III,NSC 3852,8-Hydroxy-5-nitrosoquinoline IUPAC Name: 5-nitrosoquinolin-8-ol SMILES: C1=CC2=C(C(=CC=C2N=O)O)N=C1