accessibility menu, dialog, popup

UserName

CAS RN 3529-82-6

ONONCS

CAS RN 3529-82-6

IUPAC Name: 1-isothiocyanato-3-nitrobenzene
Synonyms: 1-isothiocyanato-3-nitrobenzene
Molecular Weight (g/mol): 180.18
Molecular Formula: C7H4N2O2S
InChi Key: OEZXLKSZOAWNJU-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(=C1)N=C=S
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results