CAS RN 352457-34-2
CAS RN 352457-34-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Hydroxy Moxonidine, TRC
CAS: 352457-34-2 Molecular Formula: C9 H13 N5 O2 Molecular Weight (g/mol): 223.23 Synonym: 4(3H)-Pyrimidinone, 5-[(4,5-dihydro-1H-imidazol-2-yl)amino]-6-methoxy-2-methyl-,4(1H)-Pyrimidinone, 5-[(4,5-dihydro-1H-imidazol-2-yl)amino]-6-methoxy-2-methyl- (9CI),5-[(4,5-Dihydro-1H-imidazol-2-yl)amino]-6-methoxy-2-methyl-4(3H)-pyrimidinone,4-Hydroxymoxonidine,5-[(Imidazolidin-2-ylidene)amino]-6-methoxy-2-methylpyrimidin-4-ol IUPAC Name: 5-(imidazolidin-2-ylideneamino)-6-methoxy-2-methylpyrimidin-4-ol SMILES: COc1nc(C)nc(O)c1N=C2NCCN2