accessibility menu, dialog, popup

UserName

CAS RN 34819-86-8

CH3OOH3COOH3CO

CAS RN 34819-86-8

IUPAC Name: 3-(acetyloxy)-2-methyl-3,4-dihydro-2H-pyran-4-yl acetate
Synonyms: 3-(acetyloxy)-2-methyl-3,4-dihydro-2H-pyran-4-yl acetate
Molecular Weight (g/mol): 214.22
Molecular Formula: C10H14O5
InChi Key: NDEGMKQAZZBNBB-UHFFFAOYNA-N
SMILES: CC1OC=CC(OC(C)=O)C1OC(C)=O
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results