CAS RN 347887-36-9
CAS RN 347887-36-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Fedovapagon, MedChemExpress
MedChemExpress Fedovapagon is a selective vasopressin V2 receptor (V2R) agonist with an EC50 of 24 nM, which is being developed for the treatment of nocturia.