CAS RN 3476-89-9
CAS RN 3476-89-9
IUPAC Name:
1,2,3,4-tetrahydroquinoxaline
Synonyms:
1,2,3,4-tetrahydroquinoxaline
Molecular Weight (g/mol):
134.18
Molecular Formula:
C8H10N2
InChi Key:
HORKYAIEVBUXGM-UHFFFAOYSA-N
SMILES:
C1CNC2=CC=CC=C2N1
1
–
1
of
1
results
1
1,2,3,4-Tetrahydroquinoxaline, 98%
CAS: 3476-89-9 Molecular Formula: C8H10N2 Molecular Weight (g/mol): 134.18 MDL Number: MFCD00047564 InChI Key: HORKYAIEVBUXGM-UHFFFAOYSA-N Synonym: 1,2,3,4-tetrahydro-quinoxaline,tetrahydroquinoxaline,quinoxaline, 1,2,3,4-tetrahydro,1,3,4-tetrahydroquinoxaline,ksc495i0b,1,2,3,4-terahydroquinoxaline,quinoxaline,2,3,4-tetrahydro,1,2,3,4-tetrahydro quinoxaline,benzo 2,3 piperazine, tetrahydro PubChem CID: 77028 IUPAC Name: 1,2,3,4-tetrahydroquinoxaline SMILES: C1CNC2=CC=CC=C2N1