accessibility menu, dialog, popup

UserName

CAS RN 34612-38-9

HOOOHOOHOHOOHOOHOOHOHOOHOHOHOOHHOHO

CAS RN 34612-38-9

IUPAC Name: 2-[(6-{[4,5-dihydroxy-2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxan-3-yl]oxy}-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms: amylotetraose hydrolyzed starch α-1,4-tetraglucose
Molecular Weight (g/mol): 666.58
Molecular Formula: C24H42O21
InChi Key: LUEWUZLMQUOBSB-UHFFFAOYNA-N
SMILES: OCC1OC(OC2C(O)C(O)C(OC3C(O)C(O)C(OC4C(O)C(O)C(O)OC4CO)OC3CO)OC2CO)C(O)C(O)C1O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)