CAS RN 3422-01-3
CAS RN 3422-01-3
IUPAC Name:
tert-butyl N-phenylcarbamate
Synonyms:
tert-butyl N-phenylcarbamate
Molecular Weight (g/mol):
193.25
Molecular Formula:
C11H15NO2
InChi Key:
KZZHPWMVEVZEFG-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)NC1=CC=CC=C1
1
–
1
of
1
results
1
N-(tert-Butoxycarbonyl)aniline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.