accessibility menu, dialog, popup

UserName

CAS RN 3343-59-7

OHNHOHO(E)

CAS RN 3343-59-7

IUPAC Name: (4E)-2-hydroxy-4-[(hydroxyamino)methylidene]cyclohexa-2,5-dien-1-one
Synonyms: (4E)-2-hydroxy-4-[(hydroxyamino)methylidene]cyclohexa-2,5-dien-1-one
Molecular Weight (g/mol): 153.14
Molecular Formula: C7H7NO3
InChi Key: XQTLVFXPCMGDOB-SNAWJCMRSA-N
SMILES: ON\C=C1/C=CC(=O)C(O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results