CAS RN 332-80-9
CAS RN 332-80-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
1-Methyl-L-histidine, TRC
CAS: 332-80-9 Molecular Formula: C7 H11 N3 O2 Molecular Weight (g/mol): 169.18 Synonym: 1-Methyl-L-histidine,Histidine, 1-methyl-, L- (8CI),1-Methyl-L-histidine,(S)-2-Amino-3-(1-methyl-1H-imidazol-4-yl)propanoic acid,1-Methylhistidine,1-N-Methyl-L-histidine,N1-Methyl-L-histidine,NSC 524367 IUPAC Name: (2S)-2-amino-3-(1-methylimidazol-4-yl)propanoic acid SMILES: Cn1cnc(C[C@H](N)C(=O)O)c1
2
1-Methyl-L-histidine, MedChemExpress
MedChemExpress 1-Methyl-L-histidine is an objective indicator of meat ingestion and exogenous 3-methylhistidine (3MH) intake.