accessibility menu, dialog, popup

UserName

CAS RN 32965-49-4

ClClRhRhH2CCH2H2CCH2

CAS RN 32965-49-4

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 441
Molecular Formula: C12H20Cl2Rh2
InChi Key: GCDBRPYDOCSTAL-UHFFFAOYSA-N
SMILES: [Cl].[Cl].[Rh].[Rh].C=CCCC=C.C=CCCC=C
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results