accessibility menu, dialog, popup

UserName

CAS RN 326-66-9

CH3ONHBrF

CAS RN 326-66-9

IUPAC Name: N-(4-bromo-2-fluorophenyl)acetamide
Synonyms: N-(4-bromo-2-fluorophenyl)acetamide
Molecular Weight (g/mol): 232.05
Molecular Formula: C8H7BrFNO
InChi Key: BCYGKMDWQBWUSC-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(Br)C=C1F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results