accessibility menu, dialog, popup

UserName

CAS RN 32287-65-3

AlFFF

CAS RN 32287-65-3

IUPAC Name: trifluoroalumane hydrate
Synonyms: aluminium fluoride hydrate
Molecular Weight (g/mol): 101.99
Molecular Formula: AlF3H2O
InChi Key: XTXFDUABTPNTFB-UHFFFAOYSA-K
SMILES: O.F[Al](F)F
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results