accessibility menu, dialog, popup

UserName

CAS RN 321531-75-3

NHOClNOO

CAS RN 321531-75-3

IUPAC Name: 4-chloro-N-cyclohexyl-3-nitrobenzamide
Synonyms: 4-chloro-N-cyclohexyl-3-nitrobenzamide
Molecular Weight (g/mol): 282.72
Molecular Formula: C13H15ClN2O3
InChi Key: NXILMCJKLLFJES-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(Cl)C=CC(=C1)C(=O)NC1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results