accessibility menu, dialog, popup

UserName

CAS RN 320-97-8

HOOFOHO

CAS RN 320-97-8

IUPAC Name: 4-fluorobenzene-1,2-dicarboxylic acid
Synonyms: 4-fluorobenzene-1,2-dicarboxylic acid
Molecular Weight (g/mol): 184.12
Molecular Formula: C8H5FO4
InChi Key: OMCXTFVBNCFZMY-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(F)C=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results