accessibility menu, dialog, popup

UserName

CAS RN 31776-83-7

FNClN

CAS RN 31776-83-7

IUPAC Name: 2-chloro-6-(4-fluorophenyl)pyridine-3-carbonitrile
Synonyms: 2-chloro-6-(4-fluorophenyl)pyridine-3-carbonitrile
Molecular Weight (g/mol): 232.64
Molecular Formula: C12H6ClFN2
InChi Key: VJYNNRPTVYPCCS-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C1=CC=C(C#N)C(Cl)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results