CAS RN 3141-25-1
CAS RN 3141-25-1
IUPAC Name:
2,3,4-tribromothiophene
Synonyms:
2,3,4-tribromothiophene
Molecular Weight (g/mol):
320.82
Molecular Formula:
C4HBr3S
InChi Key:
ZDXQFDMBZUQHOM-UHFFFAOYSA-N
SMILES:
BrC1=C(Br)C(Br)=CS1
1
–
1
of
1
results
1
2,3,4-Tribromothiophene, 97%, Thermo Scientific™
CAS: 3141-25-1 Molecular Formula: C4HBr3S Molecular Weight (g/mol): 320.82 MDL Number: MFCD00041210 InChI Key: ZDXQFDMBZUQHOM-UHFFFAOYSA-N PubChem CID: 76593 IUPAC Name: 2,3,4-tribromothiophene SMILES: BrC1=C(Br)C(Br)=CS1