CAS RN 30845-11-5
CAS RN 30845-11-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
S-(3-Carboxypropyl)-L-cysteine, TRC
CAS: 30845-11-5 Molecular Formula: C7H13NO4S Molecular Weight (g/mol): 207.25 Synonym: (R)-4-[(2-Amino-2-carboxyethyl)thio]butanoic Acid,L-4-[(2-Amino-2-carboxyethyl)thio]butyric Acid,3-(3-Carboxypropylthio)alanine IUPAC Name: 4-[(2R)-2-amino-2-carboxyethyl]sulfanylbutanoic acid SMILES: C(CC(=O)O)CSC[C@@H](C(=O)O)N