accessibility menu, dialog, popup

UserName

CAS RN 308103-40-4

H3COBOHHO

CAS RN 308103-40-4

IUPAC Name: (2-acetylphenyl)boronic acid
Synonyms: 2-acetylphenylboronic acid
Molecular Weight (g/mol): 163.97
Molecular Formula: C8H9BO3
InChi Key: ZKAOVABYLXQUTI-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=CC=C1B(O)O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)