CAS RN 30544-61-7
CAS RN 30544-61-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Clanobutin, TRC
CAS: 30544-61-7 Molecular Formula: C18 H18 Cl N O4 Molecular Weight (g/mol): 347.79 Synonym: Clanobutin, 4-[(4-Chlorobenzoyl)(4-methoxyphenyl)amino]butanoic acid (ACI),Butyric acid, 4-[p-chloro-N-(p-methoxyphenyl)benzamido]- (8CI),Bykahepar,Clanobutin,Clanobutine,N-(p-Chlorobenzoyl)-γ-(p-anisidino)butyric acid IUPAC Name: 4-(N-(4-chlorobenzoyl)-4-methoxyanilino)butanoic acid SMILES: COc1ccc(cc1)N(CCCC(=O)O)C(=O)c2ccc(Cl)cc2