accessibility menu, dialog, popup

UserName

CAS RN 29927-08-0

CH3SH2NNCH3

CAS RN 29927-08-0

IUPAC Name: 5,6-dimethyl-1,3-benzothiazol-2-amine
Synonyms: 5,6-dimethyl-1,3-benzothiazol-2-amine
Molecular Weight (g/mol): 178.25
Molecular Formula: C9H10N2S
InChi Key: IODWHFFPQHUDAG-UHFFFAOYSA-N
SMILES: CC1=C(C)C=C2N=C(N)SC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)