accessibility menu, dialog, popup

UserName

CAS RN 28862-80-8

H3CS(R)OHONHOO

CAS RN 28862-80-8

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid
Synonyms: 2-{[(benzyloxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid
Molecular Weight (g/mol): 283.34
Molecular Formula: C13H17NO4S
InChi Key: FPKHNNQXKZMOJJ-UHFFFAOYNA-N
SMILES: CSCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results