accessibility menu, dialog, popup

UserName

CAS RN 283-38-5

NNH

CAS RN 283-38-5

IUPAC Name: 1,4-diazabicyclo[3.2.2]nonane
Synonyms: 1,4-diazabicyclo[3.2.2]nonane
Molecular Weight (g/mol): 126.2
Molecular Formula: C7H14N2
InChi Key: XJKNACDCUAFDHD-UHFFFAOYSA-N
SMILES: C1CN2CCC1NCC2
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results