accessibility menu, dialog, popup

UserName

CAS RN 28281-49-4

CH3OOO

CAS RN 28281-49-4

IUPAC Name: 1-(2H-1,3-benzodioxol-5-yl)propan-1-one
Synonyms: 1-(2H-1,3-benzodioxol-5-yl)propan-1-one
Molecular Weight (g/mol): 178.19
Molecular Formula: C10H10O3
InChi Key: RVBJGSPBFIUTTR-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC=C2OCOC2=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results