CAS RN 28090-12-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-(3-Methylbut-2-enoxy)benzaldehyde, TRC
CAS: 28090-12-2 Molecular Formula: C12H14O2 Molecular Weight (g/mol): 190.24 Synonym: 4-Prenyloxybenzaldehyde,4-[(3-Methyl-2-butenyl)oxy]benzaldehyde,4-[(3-Methyl-2-buten-1-yl)oxy]benzaldehyde SMILES: CC(=CCOc1ccc(C=O)cc1)C