CAS RN 2800-80-8
CAS RN 2800-80-8
IUPAC Name:
3,3-bis(3-bromo-4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Synonyms:
3,3-bis(3-bromo-4-hydroxyphenyl)-2,1λ⁶-benzoxathiole-1,1-dione
Molecular Weight (g/mol):
512.17
Molecular Formula:
C19H12Br2O5S
InChi Key:
OYCLSQDXZMROJK-UHFFFAOYSA-N
SMILES:
OC1=CC=C(C=C1Br)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(Br)=C1
1
–
2
of
2
results
1
Thermo Scientific Chemicals Bromophenol Red, pure
CAS: 2800-80-8 Molecular Formula: C19H12Br2O5S Molecular Weight (g/mol): 512.168 MDL Number: MFCD00023014 InChI Key: OYCLSQDXZMROJK-UHFFFAOYSA-N Synonym: 5', 5''-Dibromophenolsulfonphthalein PubChem CID: 76047 IUPAC Name: 2-bromo-4-[3-(3-bromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol SMILES: C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C=C3)O)Br)C4=CC(=C(C=C4)O)Br