accessibility menu, dialog, popup

UserName

CAS RN 27583-37-5

CH3HOHO

CAS RN 27583-37-5

IUPAC Name: 3-methylcyclopentane-1,2-diol
Synonyms: 3-methylcyclopentane-1,2-diol
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: KANFKJUPLALTDB-UHFFFAOYNA-N
SMILES: CC1CCC(O)C1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results