accessibility menu, dialog, popup

UserName

CAS RN 26780-50-7

OOOOMex

CAS RN 26780-50-7

IUPAC Name: 1,4-dioxane-2,5-dione; 3,6-dimethyl-1,4-dioxane-2,5-dione
Synonyms: 1,4-dioxane-2,5-dione; lactide
Molecular Weight (g/mol): 260.2
Molecular Formula: C10H12O8
InChi Key: LCSKNASZPVZHEG-UHFFFAOYNA-N
SMILES: O=C1COC(=O)CO1.CC1OC(=O)C(C)OC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results