CAS RN 264622-53-9
CAS RN 264622-53-9
IUPAC Name:
N-(4-acetylphenyl)-2-[4-(2,6-dioxo-1,3-dipropyl-2,3,6,7-tetrahydro-1H-purin-8-yl)phenoxy]acetamide
Synonyms:
N-(4-acetylphenyl)-2-[4-(2,6-dioxo-1,3-dipropyl-7H-purin-8-yl)phenoxy]acetamide
Molecular Weight (g/mol):
503.56
Molecular Formula:
C27H29N5O5
InChi Key:
ZKUCFFYOQOJLGT-UHFFFAOYSA-N
SMILES:
CCCN1C2=C(NC(=N2)C2=CC=C(OCC(=O)NC3=CC=C(C=C3)C(C)=O)C=C2)C(=O)N(CCC)C1=O
1
–
1
of
1
results
1
MRS-1706, MedChemExpress
MedChemExpress MRS-1706 is a potent and selective adenosine A2B receptor inverse agonist. MRS-1706 has Ki values of 1.39, 112, 157, and 230 nM for human A2B, A2A, A1 and A3 receptors respectively. MRS-1706 blocks adenosine-mediated cAMP induction.